Learn More
Chemglass Life Sciences Extraction Thimble, Glass, Large, X-Coarse Frit, 45mm OD X 130mm Height
SDP
Catalog No. 50873064
Encompass_Preferred
Clamp, Horseshoe Style, 5.5in Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. 50-873-064 Supplier Chemglass Life Sciences Supplier No. CG137206
Clamp, Horseshoe Style, 5.5in Flange. Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.