Learn More
Chemglass Life Sciences Extraction Thimble, Glass, Medium, Coarse Frit, 35mm OD X 90mm Height
SDP
Catalog No. 50873063
Encompass_Preferred
Extraction Thimble, Glass, Medium, Coarse Frit, 35mm OD X 90mm Height. All glass extraction thimble permits viewing during extraction process and subsequent weighing. Sizes correspond to CG-1368 or CG-1369 extractor. Coarse porosity. Sizes and porosities other than those listed are available upon request.
Catalog No. 50-873-063 Supplier Chemglass Life Sciences Supplier No. CG137203
Extraction Thimble, Glass, Medium, Coarse Frit, 35mm OD X 90mm Height. All glass extraction thimble permits viewing during extraction process and subsequent weighing. Sizes correspond to CG-1368 or CG-1369 extractor. Coarse porosity. Sizes and porosities other than those listed are available upon request.