Learn More
Chemglass Life Sciences Extraction Thimble, Glass, Small, X-Coarse Frit, 25mm OD X 85mm Height
SDP
Catalog No. 50873062
Encompass_Preferred
Extraction Thimble, Glass, Small, X-Coarse Frit, 25mm OD X 85mm Height. All glass extraction thimble permits viewing during extraction process and subsequent weighing. Sizes correspond to CG-1368 or CG-1369 extractor. Extra coarse porosity. Sizes and porosities other than those listed are available upon request.
Catalog No. 50-873-062 Supplier Chemglass Life Sciences Supplier No. CG137202
Extraction Thimble, Glass, Small, X-Coarse Frit, 25mm OD X 85mm Height. All glass extraction thimble permits viewing during extraction process and subsequent weighing. Sizes correspond to CG-1368 or CG-1369 extractor. Extra coarse porosity. Sizes and porosities other than those listed are available upon request.