Learn More
Research Products International Corp Thimerosal, 25 Grams
SDP
Catalog No. 50213684
Encompass_Preferred
Bacteriostatic and fungistatic mercurial agent.
Toxicity: LD50 98 mg/kg
WARNING! This product can expose you to Thimerosal, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-684 Supplier Research Products International Corp Supplier No. T2104025.0
Bacteriostatic and fungistatic mercurial agent.
Toxicity: LD50 98 mg/kg
WARNING! This product can expose you to Thimerosal, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.