Learn More
Electron Microscopy Sciences Gilder Thin Bar Square Combined mesh Copper Grid
SDP
Catalog No. 5028888
Encompass_Preferred
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Pitch: Copper
Catalog No. 50-288-88 Supplier Electron Microscopy Sciences Supplier No. TVMCU
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Pitch: Copper