Learn More
Electron Microscopy Sciences Gilder Thin Bar Square 1000 mesh Nickel Grid
SDP
Catalog No. 5028894
Encompass_Preferred
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 25 µm
Hole: 19 µm
Bar: 6 µm
:
Catalog No. 50-288-94 Supplier Electron Microscopy Sciences Supplier No. T1000NI
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 25 µm
Hole: 19 µm
Bar: 6 µm
:
Product Title