Learn More
Electron Microscopy Sciences Gilder Thin Bar Square 400 mesh Gold Grid
SDP
Catalog No. 5028897
Encompass_Preferred
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Gold
Pitch: 62 µm
Hole: 54 µm
Bar: 8 µm
:
Catalog No. 50-288-97 Supplier Electron Microscopy Sciences Supplier No. T400AU
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Gold
Pitch: 62 µm
Hole: 54 µm
Bar: 8 µm
:
Product Title