Learn More
Electron Microscopy Sciences Gilder Thin Bar Square 1500 mesh Nickel Grid
SDP
Catalog No. 5028348
Encompass_Preferred
Very fine mesh grids have up to 2,000 lines/inch. There is an increasing need in TEM for support films, routinely carbon, as thin as 1.5 - 2.0 nm. The pitch (the distance from the center of one bar to the center of the next bar) dimension in all grids remains constant, allowing use as a lower magnification calibration aid.
Apart from one side being shiny, the other matt, there is a large asymmetrical mark in the rim which gives the identification of which side the specimen is on. Grids are 3.05 mm overall diameter; 2.05 mm mesh area diameter.
Grid: Nickel
Pitch: 16.5 µm
Hole: 11.5 µm
Bar: 5 µm
:
Catalog No. 50-283-48 Supplier Electron Microscopy Sciences Supplier No. T1500NI
Very fine mesh grids have up to 2,000 lines/inch. There is an increasing need in TEM for support films, routinely carbon, as thin as 1.5 - 2.0 nm. The pitch (the distance from the center of one bar to the center of the next bar) dimension in all grids remains constant, allowing use as a lower magnification calibration aid.
Apart from one side being shiny, the other matt, there is a large asymmetrical mark in the rim which gives the identification of which side the specimen is on. Grids are 3.05 mm overall diameter; 2.05 mm mesh area diameter.
Grid: Nickel
Pitch: 16.5 µm
Hole: 11.5 µm
Bar: 5 µm
:
Product Title