Learn More
Electron Microscopy Sciences Gilder Thin Bar Hexagonal 200 mesh Copper Grid
SDP
Catalog No. 5026476
Encompass_Preferred
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Copper
Pitch: 125 µm
Hole: 113 µm
Bar: 12 µm
Catalog No. 50-264-76 Supplier Electron Microscopy Sciences Supplier No. T200HCU
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Copper
Pitch: 125 µm
Hole: 113 µm
Bar: 12 µm