Learn More
Electron Microscopy Sciences Gilder Thin Bar Hexagonal 300 mesh Nickel Grid
SDP
Catalog No. 5026473
Encompass_Preferred
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 83 µm
Hole: 73 µm
Bar: 10 µm
Catalog No. 50-264-73 Supplier Electron Microscopy Sciences Supplier No. T300HNI
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area. 3.05 diameter, 0.8 mil thickness.
Grid: Nickel
Pitch: 83 µm
Hole: 73 µm
Bar: 10 µm