Learn More
Bio Rad Laboratories THIN BLOT PAPER
Catalog No. 509013027
Encompass
Paper, Filter; Bio-Rad; Blot absorbent; Thin; 100% cotton; Produced using ultrapure water; 33cm x 3m; 1 roll
Catalog No. 50-901-3027 Supplier Bio Rad Laboratories Supplier No. 1620118
Paper, Filter; Bio-Rad; Blot absorbent; Thin; 100% cotton; Produced using ultrapure water; 33cm x 3m; 1 roll