Learn More
MSC AbilityOne SKILCRAFT Floor Pad, Nylon, Buffing, Tan, 21 in Diameter
Catalog No. 502839613
Encompass
These floor pads are designed for use on concentrated weight, rotary disk floor machines rated between 150 - 350 rpm. They are washable, reusable, and corrosion-resistant, suitable for various floor types including resilient tile, terrazzo, marble, and ceramic floors. Each pad measures 11/16 in thick and features a 3-3/8 in center hole.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.
Catalog No. 50-283-9613 Supplier MSC Supplier No. 89460539
These floor pads are designed for use on concentrated weight, rotary disk floor machines rated between 150 - 350 rpm. They are washable, reusable, and corrosion-resistant, suitable for various floor types including resilient tile, terrazzo, marble, and ceramic floors. Each pad measures 11/16 in thick and features a 3-3/8 in center hole.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.