Learn More
Electron Microscopy Sciences Thin Sample Holder, 15x10 mm Stub
SDP
Catalog No. 5029306
Encompass
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For most AMRAY: 1/2" diameter, 1/8" dia. Pin (3.1mm) with split openings up to 1/4" (6.4mm), with 15mm (9/16") pin height
Catalog No. 50-293-06 Supplier Electron Microscopy Sciences Supplier No. 7594810
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For most AMRAY: 1/2" diameter, 1/8" dia. Pin (3.1mm) with split openings up to 1/4" (6.4mm), with 15mm (9/16") pin height