Learn More
Electron Microscopy Sciences Thin Sample Holder, 8 mm Pin
SDP
Catalog No. 5029305
Encompass
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For most AMRAY: 1/2" diameter (12.7mm), 1/8" (3.1mm)dia. Pin (3.1mm) with split openings up to 1/4" (6.4mm), with 8mm (5/16") pin height
Catalog No. 50-293-05 Supplier Electron Microscopy Sciences Supplier No. 7594808
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For most AMRAY: 1/2" diameter (12.7mm), 1/8" (3.1mm)dia. Pin (3.1mm) with split openings up to 1/4" (6.4mm), with 8mm (5/16") pin height