Learn More
Arizona Instrument LLC Thin Septa high temperature caps 100/pk
Catalog No. NC0748642
Encompass
Septa for high temperature caps, to be used in conjunction with Computrac Vapor Pro / Vapor Pro XL moisture specific analysers. Thickness: 0.1mm, Closure Size: 24/400, 100/pk
Catalog No. NC0748642 Supplier Arizona Instrument LLC Supplier No. 9900207BX
Septa for high temperature caps, to be used in conjunction with Computrac Vapor Pro / Vapor Pro XL moisture specific analysers. Thickness: 0.1mm, Closure Size: 24/400, 100/pk