Learn More
Electron Microscopy Sciences Thin Specimen Split Mount, AMRAY, 15 mm
SDP
Catalog No. 501927135
Encompass_Preferred
Split SEM specimen holder opens to 3.75 mm. 15 mm dia with 3.2 mm pin. Pin length is 14.3 mm. Height of head: 10.2 mm, centered split. Pin is off-center. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Catalog No. 50-192-7135 Supplier Electron Microscopy Sciences Supplier No. 7595009
Split SEM specimen holder opens to 3.75 mm. 15 mm dia with 3.2 mm pin. Pin length is 14.3 mm. Height of head: 10.2 mm, centered split. Pin is off-center. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.