Learn More
Cole-Parmer Cole-Parmer elements Ultra-thin magnetic stirrer with Timer, 1L Capacity; 100-240V
Catalog No. 501960375 Shop All Cole-Parmer Products
Encompass_Preferred
Ultra flat stirrer utilizes advanced magnetic coil technology providing lighter, quieter, and wear-free operation. The unique design makes the stirrer thinner and lighter to be easily stored when not in use, saving valuable bench space. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built in timer can be set for up to 9 hours or continuous operation.
Catalog No. 50-196-0375 Supplier Cole-Parmer Supplier No. 8400371
Ultra flat stirrer utilizes advanced magnetic coil technology providing lighter, quieter, and wear-free operation. The unique design makes the stirrer thinner and lighter to be easily stored when not in use, saving valuable bench space. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built in timer can be set for up to 9 hours or continuous operation.