Learn More
Electron Microscopy Sciences Veco Thin and Thick Bars 100µK, Copper Grid
SDP
Catalog No. 50980159
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 156/132 µm
Hole: 100 µm
Bar: 56/32 µm
Catalog No. 50-980-159 Supplier Electron Microscopy Sciences Supplier No. 100SCU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 156/132 µm
Hole: 100 µm
Bar: 56/32 µm