Learn More
Electron Microscopy Sciences Veco Thin and Thick Bars GE200, Nickel Grid
SDP
Catalog No. 5029290
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 125/145 µm
Hole: 80Single slot 1000x500 µm
Bar: 45/65 µm
Catalog No. 50-292-90 Supplier Electron Microscopy Sciences Supplier No. GE200NI
Add to Cart
Add to Cart
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 125/145 µm
Hole: 80Single slot 1000x500 µm
Bar: 45/65 µm