Learn More
Sutter Instruments THIN WALL BOROSILICT GLSS,10CM
SDP
Catalog No. NC2025858
Encompass_Preferred
Thin Wall Borosilicate Glass w/Filament, 10cm, 1.50 mm OD225/pk
Catalog No. NC2025858 Supplier Sutter Instruments Supplier No. BF15011010
Thin Wall Borosilicate Glass w/Filament, 10cm, 1.50 mm OD225/pk