Learn More
Bioworld SnapStrip® PCR Tubes, Tubes Only, Assorted, 1 PK(s)
Catalog No. 50197137
Encompass_Preferred
Product Specifications:8-tube strips, 125 total strips per packTubes Only0.2 mL measurementTubes can be easily separated when desired to create smaller stripsAssorted Colors
Catalog No. 50-197-137 Supplier Bioworld Supplier No. 111900091
Product Specifications:8-tube strips, 125 total strips per packTubes Only0.2 mL measurementTubes can be easily separated when desired to create smaller stripsAssorted Colors