Learn More
MSC Excel Dryer Hand Dryer, ADA Compliant, 208-277V, Stainless Steel, 950 W
Catalog No. 502837513
Encompass
The ThinAir Hand Dryer is an ADA compliant, surface-mounted hand dryer designed for high-speed drying. It offers a 14-second dry time and operates quietly with adjustable speed and sound control. This model features a brushed stainless steel cover and variable heat settings, making it an energy-efficient and hygienic solution for various applications.
  • Surface mounted and ADA compliant
  • 14-second hand drying time
  • Adjustable speed, sound, and heat settings
  • Washable prefilter
  • Touchless, high-speed operation
  • Slim profile meets 4 inch ADA protrusion guidelines
  • Brushed stainless steel cover
  • Automatic sensor-operated activation
  • Energy efficient, with cost and carbon footprint savings
  • Reduces touch points and cross-contamination risk
  • 5-year manufacturer limited warranty
Catalog No. 50-283-7513 Supplier MSC Supplier No. 38866109
The ThinAir Hand Dryer is an ADA compliant, surface-mounted hand dryer designed for high-speed drying. It offers a 14-second dry time and operates quietly with adjustable speed and sound control. This model features a brushed stainless steel cover and variable heat settings, making it an energy-efficient and hygienic solution for various applications.
  • Surface mounted and ADA compliant
  • 14-second hand drying time
  • Adjustable speed, sound, and heat settings
  • Washable prefilter
  • Touchless, high-speed operation
  • Slim profile meets 4 inch ADA protrusion guidelines
  • Brushed stainless steel cover
  • Automatic sensor-operated activation
  • Energy efficient, with cost and carbon footprint savings
  • Reduces touch points and cross-contamination risk
  • 5-year manufacturer limited warranty