Learn More
MSC Excel Dryer Hand Dryer, White Polymer (ABS), ADA Compliant, Surface Mounted, HEPA Filtration
Catalog No. 502837755
Encompass
This ThinAir Hand Dryer is an automatic, ADA-compliant, high-speed unit with an electrostatic HEPA filter. It features a white polymer (ABS) antimicrobial cover, efficiently removing viruses and bacteria from the air while offering adjustable speed, sound, and heat settings for optimal performance and user comfort.
  • White polymer (ABS) antimicrobial cover
  • Electrostatic HEPA filtration system removes 99.999% of viruses and 99.97% of bacteria
  • ADA compliant, surface-mounted design
  • 14-second dry time
  • Adjustable speed, sound, and heat
  • Washable metal mesh pre-filter
  • Up to 95% cost savings versus paper towels
  • 5-year parts warranty
Catalog No. 50-283-7755 Supplier MSC Supplier No. 63493415
This ThinAir Hand Dryer is an automatic, ADA-compliant, high-speed unit with an electrostatic HEPA filter. It features a white polymer (ABS) antimicrobial cover, efficiently removing viruses and bacteria from the air while offering adjustable speed, sound, and heat settings for optimal performance and user comfort.
  • White polymer (ABS) antimicrobial cover
  • Electrostatic HEPA filtration system removes 99.999% of viruses and 99.97% of bacteria
  • ADA compliant, surface-mounted design
  • 14-second dry time
  • Adjustable speed, sound, and heat
  • Washable metal mesh pre-filter
  • Up to 95% cost savings versus paper towels
  • 5-year parts warranty