Learn More
MSC Excel Dryer ThinAir Hand Dryer, Graphite, 110-120V, eHEPA
Catalog No. 502837756
Encompass
This ThinAir hand dryer is a surface-mounted, ADA compliant unit featuring eHEPA filtration for enhanced hygiene. It delivers a rapid 14-second dry time and offers substantial cost and energy savings compared to traditional paper towels, providing an efficient and sanitary hand drying solution for various environments.
  • Automatic sensor activation for touchless operation.
  • Graphite epoxy-painted galvanized steel construction.
  • Meets 4 inch ADA protrusion guidelines.
  • eHEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • Achieves 95% cost savings versus paper towels.
  • Consumes 80% less energy than conventional hand dryers.
  • Adjustable heat, speed, and sound settings.
  • Backed by a 5-year limited warranty.
Catalog No. 50-283-7756 Supplier MSC Supplier No. 63493449
This ThinAir hand dryer is a surface-mounted, ADA compliant unit featuring eHEPA filtration for enhanced hygiene. It delivers a rapid 14-second dry time and offers substantial cost and energy savings compared to traditional paper towels, providing an efficient and sanitary hand drying solution for various environments.
  • Automatic sensor activation for touchless operation.
  • Graphite epoxy-painted galvanized steel construction.
  • Meets 4 inch ADA protrusion guidelines.
  • eHEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • Achieves 95% cost savings versus paper towels.
  • Consumes 80% less energy than conventional hand dryers.
  • Adjustable heat, speed, and sound settings.
  • Backed by a 5-year limited warranty.