Learn More
MSC Excel Dryer Hand Dryer, ThinAir, eHEPA Filtration, Brushed Stainless, 110-120V
Catalog No. 502837827
Encompass
The ThinAir Hand Dryer offers an affordable, slim, surface-mounted solution that meets ADA compliance. Delivering a rapid 14-second dry time, it provides significant cost savings over paper towels and reduces carbon footprint. Equipped with an electrostatic HEPA (eHEPA) filtration system, it enhances hygiene by removing viruses and bacteria from the airstream.
  • Surface mounted and ADA compliant.
  • Up to 95% cost savings versus paper towels.
  • eHEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • 14-second dry time.
  • Adjustable speed, sound, and heat settings.
  • Multi-voltage operation.
  • Washable pre-filter.
  • Brushed stainless steel cover.
  • 5-year limited warranty.
Catalog No. 50-283-7827 Supplier MSC Supplier No. 63493357
The ThinAir Hand Dryer offers an affordable, slim, surface-mounted solution that meets ADA compliance. Delivering a rapid 14-second dry time, it provides significant cost savings over paper towels and reduces carbon footprint. Equipped with an electrostatic HEPA (eHEPA) filtration system, it enhances hygiene by removing viruses and bacteria from the airstream.
  • Surface mounted and ADA compliant.
  • Up to 95% cost savings versus paper towels.
  • eHEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • 14-second dry time.
  • Adjustable speed, sound, and heat settings.
  • Multi-voltage operation.
  • Washable pre-filter.
  • Brushed stainless steel cover.
  • 5-year limited warranty.