Learn More
MSC Excel Dryer Hand Dryer, White Epoxy Painted Steel, 110-120V, eHEPA Filtration
Catalog No. 502837053
Encompass
The Excel Dryer ThinAir Hand Dryer is designed for high-speed drying with energy efficiency. It features a durable white epoxy painted steel cover and operates on 110-120V. This model includes eHEPA filtration for improved hygiene and meets ADA protrusion guidelines, making it suitable for various commercial and public restrooms.
  • 14-second dry time.
  • Meets 4 in ADA protrusion guidelines when surface mounted.
  • Electrostatic HEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • 95% cost savings versus paper towels.
  • Touchless activation for hygienic operation.
  • Adjustable speed, sound, and heat control.
  • 5-year limited warranty.
  • Washable metal mesh pre-filter.
  • Building Green approved and contributes to WELL points & LEED credits.
Catalog No. 50-283-7053 Supplier MSC Supplier No. 63493134
The Excel Dryer ThinAir Hand Dryer is designed for high-speed drying with energy efficiency. It features a durable white epoxy painted steel cover and operates on 110-120V. This model includes eHEPA filtration for improved hygiene and meets ADA protrusion guidelines, making it suitable for various commercial and public restrooms.
  • 14-second dry time.
  • Meets 4 in ADA protrusion guidelines when surface mounted.
  • Electrostatic HEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
  • 95% cost savings versus paper towels.
  • Touchless activation for hygienic operation.
  • Adjustable speed, sound, and heat control.
  • 5-year limited warranty.
  • Washable metal mesh pre-filter.
  • Building Green approved and contributes to WELL points & LEED credits.