Learn More
USA Scientific Inc ThinCert 0.4 micron transp
Catalog No. 502745192
Encompass
ThinCert 0.4 um transparent insert for 6-well plates, individually wrapped, 24/pack. Includes 4 plates with lids.
Catalog No. 50-274-5192 Supplier USA Scientific Inc Supplier No. 56657641
ThinCert 0.4 um transparent insert for 6-well plates, individually wrapped, 24/pack. Includes 4 plates with lids.