Learn More
USA Scientific Inc ThinCert 1 micron trans insert
Catalog No. 502744944
Encompass
ThinCert 1 um transparent insert for 6-well plates, individually wrapped, 24/pack. Includes 4 plates with lids.
Catalog No. 50-274-4944 Supplier USA Scientific Inc Supplier No. 56657610
ThinCert 1 um transparent insert for 6-well plates, individually wrapped, 24/pack. Includes 4 plates with lids.