Learn More
USA Scientific Inc ThinCert 3 micron trans insert
Supplier:  USA Scientific Inc 56657630
Encompass
ThinCert 3 um transparent insert for 6-well plates, individually wrapped, 24/pack. Includes 4 plates with lids.
Catalog No. 50-274-5052