Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Thinline Biopsy Cassettes in Tube Packs

Catalog No. B851737WH Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851737WH White
B851737BE Beige
B851737BL Blue
B851737GN Green
B851737GY Gray
B851737LA Lavender
B851737LG Light Green
B851737LI Lilac
B851737OR Orange
B851737PK Pink
B851737TN Tan
B851737YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. B851737WH Supplier Epredia™ Supplier No. B851737WH
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biopsy tissue samples, cassettes have a front lid hinge, attached lids in matching colors, and are compatible with all Epredia PrintMate™ Cassette Printers

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Small Squares
Quantity 750/Cs.
Closure Included Yes
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color White
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Thinline Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.