Learn More
TARGETMOL CHEMICALS INC Thio-TEPA 50MG
SDP
Catalog No. 502875926
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Thio-TEPA (Tiofosyl) is a polyfunctional, organophosphorus alkylating agent with antineoplastic activity. Purity 98.45%
Catalog No. 50-287-5926 Supplier TARGETMOL CHEMICALS INC Supplier No. T125050MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Thio-TEPA (Tiofosyl) is a polyfunctional, organophosphorus alkylating agent with antineoplastic activity. Purity 98.45%