Learn More
Medchemexpress LLC Thio-TEPA | 52-24-4 | 100.0% | 189.22 | 25 MG
SDP
Catalog No. 5000384002
Encompass_Preferred
Thio-TEPA is a DNA alkylating agent with antitumor activity. It exhibits alkylating activity in rat liver slice incubation and has been shown to improve both short- and long-term engraftment in mice when administered alone.
  • DNA alkylating agent
  • Exhibits antitumor activity
  • Exhibits alkylating activity in rat liver slice incubation
  • Improves both short- and long-term engraftment in mice
Catalog No. 50-003-84002 Supplier Medchemexpress LLC Supplier No. 25MGHY17574
Thio-TEPA is a DNA alkylating agent with antitumor activity. It exhibits alkylating activity in rat liver slice incubation and has been shown to improve both short- and long-term engraftment in mice when administered alone.
  • DNA alkylating agent
  • Exhibits antitumor activity
  • Exhibits alkylating activity in rat liver slice incubation
  • Improves both short- and long-term engraftment in mice