Learn More
Medchemexpress LLC Thio-TEPA | 52-24-4 | 99.9% | 189.22 | 100 MG
SDP
Catalog No. 5000387521
Encompass_Preferred
Thio-TEPA is a DNA alkylating agent with antitumor activity. It is for research use only and not sold to patients. It exhibits alkylating activity in rat liver slice incubation and does not affect the viability of rat liver slices.
  • Exhibits alkylating activity in rat liver slice incubation.
  • Enhances donor-type blood chimerism when administered with total body irradiation (TBI).
  • Improves both short- and long-term engraftment in mice when administered alone.
Catalog No. 50-003-87521 Supplier Medchemexpress LLC Supplier No. HY17574100MG
Thio-TEPA is a DNA alkylating agent with antitumor activity. It is for research use only and not sold to patients. It exhibits alkylating activity in rat liver slice incubation and does not affect the viability of rat liver slices.
  • Exhibits alkylating activity in rat liver slice incubation.
  • Enhances donor-type blood chimerism when administered with total body irradiation (TBI).
  • Improves both short- and long-term engraftment in mice when administered alone.