Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide ACS AR, Macron Fine Chemicals™

Catalog No. MK1864100 Shop All Avantor Products
Change view
Click to view available options
Glass; 12 x 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-1864-100 Glass; 12 x 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-1864-100 Supplier Avantor Supplier No. 186412

Specifications

CAS 62-55-5
Molecular Formula C2H5NS
MDL Number MFCD00008070
Packaging Glass
Quantity 12 x 100 g
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILES CC(N)=S
IUPAC Name ethanethioamide
Molecular Weight (g/mol) 75.13
PubChem CID 2723949
ChEBI CHEBI:32497
Formula Weight 75.13
Grade ACS
Chemical Name or Material Thioacetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.