Learn More
Cayman Chemical Thioacetazone, Purity: ≥98%, CAS: 104-06-3, 100 mg
SDP
Catalog No. 502949584
Encompass_Preferred
An antitubercular agent; active against M. tuberculosis H37Rv, M. kansasii Bostrom, and M. avium serotype 2 (MICs = 0.3, 0.6, and 20 µg/ml, respectively)
Catalog No. 50-294-9584 Supplier Cayman Chemical Supplier No. 34589100 MG
An antitubercular agent; active against M. tuberculosis H37Rv, M. kansasii Bostrom, and M. avium serotype 2 (MICs = 0.3, 0.6, and 20 µg/ml, respectively)