Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thioacetic Acid

Catalog No. T30805500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30805500G 500 g
T30805100G 100 g
T308055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. T30805500G Supplier Sigma Aldrich Supplier No. T30805500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thioacetic acid is a reagent for introduction of the thiol group into organic molecules.

Specifications

CAS 507-09-5
Density 1.065 g/mL at 25°C
Boiling Point 88°C to 91.5°C
Molecular Formula C2H4OS
Linear Formula CH3COSH
Refractive Index n20/D 1.465
MDL Number MFCD00004853
Quantity 500 g
Synonym TAA; TMA; Thiacetic acid
Molecular Weight (g/mol) 76.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.