Learn More
Cayman Chemical ThIoargInInhydrobromIde 10mg
SDP
Catalog No. 502895167
Encompass_Preferred
A colorimetric arginase substrate; hydrolyzed to 2-amino-5-mercaptovaleric acid, which reacts with DTNB to form 2-nitro-5-thiobenzoate and can be quantified by colorimetric detection at 412 nm as a measure of arginase activity
Catalog No. 50-289-5167 Supplier Cayman Chemical Supplier No. 8024010 MG
A colorimetric arginase substrate; hydrolyzed to 2-amino-5-mercaptovaleric acid, which reacts with DTNB to form 2-nitro-5-thiobenzoate and can be quantified by colorimetric detection at 412 nm as a measure of arginase activity