Learn More
ATCC Lactobacillus jensenii; genomic DNA
Catalog No. 502381271
Encompass
Genomic DNA isolated from Lactobacillus jensenii strain 62G. This bacterial strain is also available as ATCC Catalog No. 25258™.
Catalog No. 50-238-1271 Supplier ATCC Supplier No. 25258D5
Genomic DNA isolated from Lactobacillus jensenii strain 62G. This bacterial strain is also available as ATCC Catalog No. 25258™.