Learn More
Medchemexpress LLC Thiobis-β-galactose-propyne | 1989627-40-8 | 98.0% | C27H40O12S2 | 10MG
SDP
Catalog No. 5000258400
Encompass_Preferred
Thiobis-β-galactose-propyne is a thiobis-linked galactose derivative used as a multivalent galectin inhibitor in preclinical research. It is supplied as a solid and intended for biochemical and cell-based studies investigating galectin-mediated interactions.
  • Multivalent galectin inhibitor suitable for biochemical assays.
  • Contains a propyne functional group for click-chemistry conjugation.
  • Reported high purity appropriate for in vitro applications.
  • Supplied as a solid and stable under recommended frozen storage.
  • Available in small mass vials convenient for screening and conjugation work.
Catalog No. 50-002-58400 Supplier Medchemexpress LLC Supplier No. HY14431210MG
Thiobis-β-galactose-propyne is a thiobis-linked galactose derivative used as a multivalent galectin inhibitor in preclinical research. It is supplied as a solid and intended for biochemical and cell-based studies investigating galectin-mediated interactions.
  • Multivalent galectin inhibitor suitable for biochemical assays.
  • Contains a propyne functional group for click-chemistry conjugation.
  • Reported high purity appropriate for in vitro applications.
  • Supplied as a solid and stable under recommended frozen storage.
  • Available in small mass vials convenient for screening and conjugation work.