Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiochrome, MilliporeSigma™ Supelco™

Catalog No. 111015909 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5909 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5909 Supplier MilliporeSigma Supelco Supplier No. T7891100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 92-35-3
Physical Form Neat
Linear Formula C12H14N4 OS
MDL Number MFCD00010519
Synonym 2,7-Dimethylthiachromine-8-ethanol
Type Analytical Standard
Quantity 100 mg
Formula Weight 262.33
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.