Learn More
Cayman Chemical ThIocolchIcIn 100mg
SDP
Catalog No. 502966669
Encompass_Preferred
An inhibitor of microtubule assembly (IC50 = 2.5 µM); inhibits binding of colchicine to tubulin (Ki = 0.7 µM); inhibits proliferation of MDA-MB-231 and multidrug resistant (MDR) MCF-7 ADRr breast cancer cells (IC50s = 0.6 and 400 nM, respectively) as well MDR CEM-VBL leukemia cells (IC50 = 50 nM)
Catalog No. 50-296-6669 Supplier Cayman Chemical Supplier No. 17427100 MG
An inhibitor of microtubule assembly (IC50 = 2.5 µM); inhibits binding of colchicine to tubulin (Ki = 0.7 µM); inhibits proliferation of MDA-MB-231 and multidrug resistant (MDR) MCF-7 ADRr breast cancer cells (IC50s = 0.6 and 400 nM, respectively) as well MDR CEM-VBL leukemia cells (IC50 = 50 nM)