Learn More
Medchemexpress LLC Thiocolchicine | 100MG
SDP
Catalog No. 5000226418
Encompass_Preferred
Thiocolchicine a derivative modified in the C Ring of Colchicine (HY-16569) with enhanced biological properties Thiocolchicine is a potent inhibitor of tubulin polymerization (IC50 2 5 M) and competitively binds to tubulin with a Ki of 0 7 M Thiocolchicine induces cell apoptosis[1][2] Thiocolchicine can be used as an ADC cytotoxin in ADC technology
Catalog No. 50-002-26418 Supplier Medchemexpress LLC Supplier No. HY116852100MG
Thiocolchicine a derivative modified in the C Ring of Colchicine (HY-16569) with enhanced biological properties Thiocolchicine is a potent inhibitor of tubulin polymerization (IC50 2 5 M) and competitively binds to tubulin with a Ki of 0 7 M Thiocolchicine induces cell apoptosis[1][2] Thiocolchicine can be used as an ADC cytotoxin in ADC technology