Learn More
AdipoGen Thiocolchicine (25 mg)
SDP
Supplier:  AdipoGen AGCN20074M025
Encompass_Preferred
Chemical. CAS: 2730-71-4. Formula: C22H25NO5S. MW: 415.5. Semisynthetic. Antimitotic alkaloid. Tubulin polymerization and microtubule assembly inhibitor. Axonal cytoskeleton modulator. Inhibitor of autophagosome-lysosome fusion. Apoptosis inducer. Thiocolchicine-dimers were shown to be potent topoisomerase I inhibitors.
Catalog No. 50-205-3426