Learn More
Medchemexpress LLC Thiocolchicine-d3 1mg | 1314417-95-2 | 418.52 | C22H22D3NO5S | 1 MG
SDP
Catalog No. 5000226210
Encompass_Preferred
Thiocolchicine-d3 is a deuterium-labeled derivative of thiocolchicine used for research applications. It inhibits tubulin polymerization (reported IC50 = 2.5 μM; Ki = 0.7 μM), induces apoptosis, and can be used as a tracer or internal standard for quantitative NMR, GC-MS, or LC-MS.
  • Deuterium-labeled tracer/internal standard for analytical quantitation.
  • Tubulin polymerization inhibitor with reported IC50 = 2.5 μM.
  • Reported Ki for tubulin binding of 0.7 μM.
  • High purity (≥99.0%).
  • Small pack sizes suitable for analytical applications.
  • Store protected from light; stable at -80°C in solvent for up to 6 months.
Catalog No. 50-002-26210 Supplier Medchemexpress LLC Supplier No. HY116852S1MG
Thiocolchicine-d3 is a deuterium-labeled derivative of thiocolchicine used for research applications. It inhibits tubulin polymerization (reported IC50 = 2.5 μM; Ki = 0.7 μM), induces apoptosis, and can be used as a tracer or internal standard for quantitative NMR, GC-MS, or LC-MS.
  • Deuterium-labeled tracer/internal standard for analytical quantitation.
  • Tubulin polymerization inhibitor with reported IC50 = 2.5 μM.
  • Reported Ki for tubulin binding of 0.7 μM.
  • High purity (≥99.0%).
  • Small pack sizes suitable for analytical applications.
  • Store protected from light; stable at -80°C in solvent for up to 6 months.