Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiocolchicine-d3 5mg | 1314417-95-2 | 418.52 | C22H22D3NO5S | 5 MG
SDP

Catalog No. 5000228957
Encompass_Preferred

Thiocolchicine-d3 is the deuterated analogue of thiocolchicine, a colchicine-site tubulin polymerization inhibitor provided for laboratory research use. It is supplied as a high-purity solid for analytical, tracer, and mechanistic studies.

  • Research use only, not for human or veterinary use.
  • Deuterated (d3) form for isotopic labeling and tracer experiments.
  • CAS 1314417-95-2 for unambiguous identification.
  • Purity typically reported at 99.0%.
  • Chemical formula C22H22D3NO5S and molecular weight ~418.52 g/mol.
  • SDS and COA available from the manufacturer product page.

Catalog No. 50-002-28957 Supplier Medchemexpress LLC Supplier No. HY116852S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiocolchicine-d3 is the deuterated analogue of thiocolchicine, a colchicine-site tubulin polymerization inhibitor provided for laboratory research use. It is supplied as a high-purity solid for analytical, tracer, and mechanistic studies.

  • Research use only, not for human or veterinary use.
  • Deuterated (d3) form for isotopic labeling and tracer experiments.
  • CAS 1314417-95-2 for unambiguous identification.
  • Purity typically reported at 99.0%.
  • Chemical formula C22H22D3NO5S and molecular weight ~418.52 g/mol.
  • SDS and COA available from the manufacturer product page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.