Learn More
AdipoGen Thiocolchicoside
SDP
Catalog No. 502559971
Encompass_Preferred
Chemical. CAS 602-41-5. Formula C27H33NO10S. MW 563.6. Synthetic. Potent competitive gamma-aminobutyric acid type A GABAA receptor antagonist and glycine receptor agonist. Described to be a muscle relaxant. Anti-inflammatory agent with analgesic properties. Shown to be an anticancer compound through inhibition of NF-kappaB and NF-kappaB-regulated gene products. Therapeutic option for the management of bone metastatic disease.
Catalog No. 50-255-9971 Supplier AdipoGen Supplier No. CDXT0175M100
Chemical. CAS 602-41-5. Formula C27H33NO10S. MW 563.6. Synthetic. Potent competitive gamma-aminobutyric acid type A GABAA receptor antagonist and glycine receptor agonist. Described to be a muscle relaxant. Anti-inflammatory agent with analgesic properties. Shown to be an anticancer compound through inhibition of NF-kappaB and NF-kappaB-regulated gene products. Therapeutic option for the management of bone metastatic disease.