Learn More
Cayman Chemical ThIocolchIcosIde 10mg
SDP
Catalog No. 502912718
Encompass_Preferred
A GABAA receptor antagonist; inhibits GABAA receptor mediated currents in Purkinje cells isolated from rat cerebellum (IC50 = 0.15 µl); binds strychnine-sensitive glycine receptors in isolated rat spinal cord (IC50 = 1.58 µM); inhibits cell growth in a panel of cancer cell lines at 25-100 µM; inhibits anti-apoptotic proteins Bcl-2, XIAP, Mcl-1, Bcl-xL, cIAP-1, and cIAP-2 in KBM5 cells; inhibits TNF-α-induced NF-κB activation and IκBα degradation in KBM5 cells
Catalog No. 50-291-2718 Supplier Cayman Chemical Supplier No. 3582110 MG
A GABAA receptor antagonist; inhibits GABAA receptor mediated currents in Purkinje cells isolated from rat cerebellum (IC50 = 0.15 µl); binds strychnine-sensitive glycine receptors in isolated rat spinal cord (IC50 = 1.58 µM); inhibits cell growth in a panel of cancer cell lines at 25-100 µM; inhibits anti-apoptotic proteins Bcl-2, XIAP, Mcl-1, Bcl-xL, cIAP-1, and cIAP-2 in KBM5 cells; inhibits TNF-α-induced NF-κB activation and IκBα degradation in KBM5 cells