Learn More
AdipoGen Thiocolchicoside (25 mg)
SDP
Supplier:  AdipoGen AGCN20076M025
Encompass_Preferred
Chemical. CAS: 602-41-5. Formula: C27H33NO10S. MW: 563.6. Semisynthetic. Potent competitive gamma-aminobutyric acid type A (GABAA) receptor antagonist and glycine receptor agonist. Weak nicotinic acetylcholine receptor agonist. Muscle relaxant. Anti-inflammatory. Has analgesic properties. Shows strong epileptogenic and convulsant activity. Anticancer compound through inhibition of NF-kappaB and NF-kappaB-regulated gene products Apoptosis inducer. Suppressed osteoclastogenesis induced by RANKL and tumor cells via the NF-kappaB signaling pathway. Therapeutic option for the management of bone metastatic disease.
Catalog No. 50-205-3431