Learn More
Selleck Chemical LLC Thioctic acid S3996-1g
SDP
Catalog No. 503109644
Encompass_Preferred
Thioctic acid (DL- -Lipoic acid) is a cyclic disulfide antioxidant that interconverts with its reduced dithiol form
Catalog No. 50-310-9644 Supplier Selleck Chemical LLC Supplier No. S39961G
Thioctic acid (DL- -Lipoic acid) is a cyclic disulfide antioxidant that interconverts with its reduced dithiol form