Learn More
Thiocyanate in Water, WP Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111004728 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-100-4728 2 mL
1 options
Catalog No. 11-100-4728 Supplier MilliporeSigma Supelco Supplier No. PE11492ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This PT sample is packaged in a 2 mL ampule as a concentrate. The solvent for this sample is water. The PT sample has not been preserved. It is recommended to prepare and analyze immediately after opening the ampule. The sample must be stored at 4°C. Sample Preparation: Partially fill a 1000-mL volumetric flask with reagent-grade water (near 20°C). Open ampule PEI-068 (be careful to avoid cuts on glass) and transfer 1.0 mL of concentrate to the volumetric flask. Fill the flask to volume with reagent-grade water and mix well. The sample is now ready for immediate analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) Thiocyanate 1-10 mg/L